Assay Details
| Gene Symbol: smarcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007296.1 | NP_001007297.1 | 8 - 9 | 1129 | 96 | |
| XM_005167183.4 | XP_005167240.1 | 8 - 9 | 1144 | 96 | ||
| GenBank mRNA | BC085668.1 | - | 8 - 9 | 1129 | 96 | |
| BC154619.1 | - | 8 - 9 | 1140 | 96 | ||
More Information
Set Membership: |
|

smarcb1a
Genomic Map