Assay Details
| Gene Symbol: sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007361.1 | NP_001007362.1 | 7 - 8 | 481 | 77 | |
| XM_021473696.1 | XP_021329371.1 | 6 - 7 | 421 | 77 | ||
| GenBank mRNA | BC085582.1 | - | 7 - 8 | 481 | 77 | |
| BC152151.1 | - | 7 - 8 | 481 | 77 | ||
| BC164350.1 | - | 7 - 8 | 411 | 77 | ||
More Information
Set Membership: |
|

sugt1
Genomic Map