Assay Details
| Gene Symbol: gorasp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007411.1 | NP_001007412.1 | 6 - 7 | 770 | 70 | |
| GenBank mRNA | BC085376.1 | - | 6 - 7 | 770 | 70 | |
| BC165574.1 | - | 6 - 7 | 681 | 70 | ||
More Information
Set Membership: |
|

gorasp1a
Genomic Map