Assay Details
| Gene Symbol: gorasp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007411.1 | NP_001007412.1 | 7 - 8 | 855 | 68 | |
| XM_005171230.4 | XP_005171287.1 | 7 - 8 | 929 | 68 | ||
| GenBank mRNA | BC085376.1 | - | 7 - 8 | 855 | 68 | |
| BC165574.1 | - | 7 - 8 | 766 | 68 | ||
More Information
Set Membership: |
|

gorasp1a
Genomic Map