Assay Details
| Gene Symbol: crygs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012390.1 | NP_001012390.1 | 2 - 3 | 289 | 83 | |
| GenBank mRNA | AY738748.1 | - | 2 - 3 | 289 | 83 | |
| BC093207.1 | - | 728 | 83 | |||
| BC164979.1 | - | 2 - 3 | 325 | 83 | ||
More Information
Set Membership: |
|

crygs3
Genomic Map