Assay Details
| Gene Symbol: rab6bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013467.1 | NP_001013485.1 | 6 - 7 | 641 | 69 | |
| GenBank mRNA | BC091529.1 | - | 6 - 7 | 641 | 69 | |
More Information
Set Membership: |
|

rab6bb
Genomic Map