Assay Details
| Gene Symbol: cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013506.1 | NP_001013524.1 | 1 - 2 | 339 | 88 | |
| GenBank mRNA | BC090428.1 | - | 1 - 2 | 339 | 88 | |
| BC165858.1 | - | 1 - 2 | 359 | 88 | ||
More Information
Set Membership: |
|

cog8
Genomic Map