Assay Details
| Gene Symbol: arsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013543.2 | NP_001013561.2 | 6 - 7 | 1278 | 112 | |
| GenBank mRNA | BC090818.1 | - | 6 - 7 | 1278 | 112 | |
More Information
Set Membership: |
|

arsa
Genomic Map