Assay Details
| Gene Symbol: trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014309.2 | NP_001014331.1 | 6 - 7 | 496 | 102 | |
| GenBank mRNA | BC091859.1 | - | 6 - 7 | 500 | 102 | |
More Information
Set Membership: |
|

trmt13
Genomic Map