Assay Details
| Gene Symbol: kcnab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014354.1 | NP_001014376.1 | 8 - 9 | 916 | 70 | |
| XM_021467646.1 | XP_021323321.1 | 8 - 9 | 826 | 70 | ||
| XM_021467647.1 | XP_021323322.1 | 8 - 9 | 824 | 70 | ||
| XM_021467648.1 | XP_021323323.1 | 8 - 9 | 871 | 70 | ||
| XM_021467649.1 | XP_021323324.1 | 8 - 9 | 871 | 70 | ||
| XM_021467650.1 | XP_021323325.1 | 8 - 9 | 1280 | 70 | ||
| GenBank mRNA | BC091978.1 | - | 8 - 9 | 916 | 70 | |
| BC096912.1 | - | 7 - 8 | 450 | 70 | ||
More Information
Set Membership: |
|

kcnab1a
Genomic Map