Assay Details
| Gene Symbol: sema3ga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014823.1 | NP_001014823.1 | 3 - 4 | 338 | 82 | |
| XM_005167031.4 | XP_005167088.1 | 4 - 5 | 973 | 82 | ||
| GenBank mRNA | AY766120.1 | - | 3 - 4 | 338 | 82 | |
| BC163710.1 | - | 3 - 4 | 422 | 82 | ||
| BC163715.1 | - | 3 - 4 | 422 | 82 | ||
More Information
Set Membership: |
|

sema3ga
Genomic Map