Assay Details
| Gene Symbol: fam8a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017551.2 | NP_001017551.1 | 3 - 4 | 721 | 103 | |
| GenBank mRNA | BC092164.1 | - | 3 - 4 | 725 | 103 | |
| BC129283.1 | - | 3 - 4 | 761 | 103 | ||
More Information
Set Membership: |
|

fam8a1b
Genomic Map