Assay Details
| Gene Symbol: fam8a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017551.2 | NP_001017551.1 | 4 - 5 | 853 | 85 | |
| GenBank mRNA | BC092164.1 | - | 4 - 5 | 857 | 85 | |
| BC129283.1 | - | 4 - 5 | 893 | 85 | ||
More Information
Set Membership: |
|

fam8a1b
Genomic Map