Assay Details
| Gene Symbol: golga7ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017566.2 | NP_001017566.1 | 2 - 3 | 256 | 88 | |
| XM_005160170.4 | XP_005160227.1 | 2 - 3 | 433 | 88 | ||
| GenBank mRNA | BC080249.1 | - | 2 - 3 | 436 | 88 | |
| BC092359.1 | - | 2 - 3 | 260 | 88 | ||
More Information
Set Membership: |
|

golga7ba
Genomic Map