Assay Details
| Gene Symbol: cdk5r2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017569.1 | NP_001017569.1 | 1 | 1044 | 64 | |
| GenBank mRNA | BC092745.1 | - | 1 | 1044 | 64 | |
More Information
Set Membership: |
|

cdk5r2a
Genomic Map