Assay Details
| Gene Symbol: zgc:165532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351801.1 | NP_001338730.1 | 7 - 8 | 891 | 88 | |
| GenBank mRNA | BC150228.1 | - | 7 - 8 | 847 | 88 | |
| EH552741.1 | - | 7 - 8 | 649 | 88 | ||
| EH573865.1 | - | 2 - 3 | 463 | 88 | ||
More Information
Set Membership: |
|

zgc:165532
Genomic Map