Assay Details
| Gene Symbol: zgc:165532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351801.1 | NP_001338730.1 | 8 - 9 | 1005 | 140 | |
| GenBank mRNA | BC150228.1 | - | 8 - 9 | 961 | 140 | |
| EH573865.1 | - | 1 - 2 | 349 | 140 | ||
More Information
Set Membership: |
|

zgc:165532
Genomic Map