Assay Details
| Gene Symbol: ap1s3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017720.1 | NP_001017720.1 | 2 - 3 | 256 | 126 | |
| XM_009291950.3 | XP_009290225.1 | 2 - 3 | 249 | 126 | ||
| XM_009291951.3 | XP_009290226.1 | 2 - 3 | 249 | 126 | ||
| XM_021481407.1 | XP_021337082.1 | 2 - 3 | 359 | 126 | ||
| GenBank mRNA | BC093241.1 | - | 2 - 3 | 256 | 126 | |
More Information
Set Membership: |
|

ap1s3a
Genomic Map