Assay Details
| Gene Symbol: ap1s3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017720.1 | NP_001017720.1 | 4 - 5 | 501 | 102 | |
| XM_009291950.3 | XP_009290225.1 | 4 - 5 | 494 | 102 | ||
| GenBank mRNA | BC093241.1 | - | 4 - 5 | 501 | 102 | |
More Information
Set Membership: |
|

ap1s3a
Genomic Map