Assay Details
| Gene Symbol: esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017796.2 | NP_001017796.1 | 7 - 8 | 743 | 115 | |
| GenBank mRNA | BC093144.1 | - | 7 - 8 | 656 | 115 | |
| BC165876.1 | - | 6 - 7 | 638 | 115 | ||
| EH478333.1 | - | 2 - 3 | 458 | 115 | ||
| EH605044.1 | - | 2 - 3 | 462 | 115 | ||
More Information
Set Membership: |
|

esd
Genomic Map