Assay Details
| Gene Symbol: creb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017818.1 | NP_001017818.1 | 4 - 5 | 456 | 106 | |
| XM_005167700.4 | XP_005167757.1 | 4 - 5 | 500 | 106 | ||
| XM_005167701.4 | XP_005167758.1 | 4 - 5 | 657 | 106 | ||
| GenBank mRNA | BC092799.1 | - | 4 - 5 | 456 | 106 | |
| BC164423.1 | - | 2 - 3 | 285 | 106 | ||
More Information
Set Membership: |
|

creb1b
Genomic Map