Assay Details
| Gene Symbol: creb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017818.1 | NP_001017818.1 | 5 - 6 | 554 | 87 | |
| XM_005167700.4 | XP_005167757.1 | 5 - 6 | 598 | 87 | ||
| XM_005167701.4 | XP_005167758.1 | 5 - 6 | 755 | 87 | ||
| GenBank mRNA | BC092799.1 | - | 5 - 6 | 554 | 87 | |
| BC164423.1 | - | 3 - 4 | 383 | 87 | ||
More Information
Set Membership: |
|

creb1b
Genomic Map