Assay Details
| Gene Symbol: dnmt3aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018134.1 | NP_001018144.1 | 18 - 19 | 2263 | 81 | |
| XM_005160858.4 | XP_005160915.1 | 21 - 22 | 3158 | 81 | ||
| GenBank mRNA | AB196919.1 | - | 18 - 19 | 2263 | 81 | |
| BC162629.1 | - | 17 - 18 | 2260 | 81 | ||
More Information
Set Membership: |
|

dnmt3aa
Genomic Map