Assay Details
| Gene Symbol: gtf2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020605.1 | NP_001018441.1 | 2 - 3 | 277 | 70 | |
| GenBank mRNA | BC095322.1 | - | 2 - 3 | 277 | 70 | |
| BC165482.1 | - | 2 - 3 | 208 | 70 | ||
More Information
Set Membership: |
|

gtf2a2
Genomic Map