Assay Details
| Gene Symbol: cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020660.1 | NP_001018496.1 | 13 - 14 | 1364 | 64 | |
| XM_005173452.4 | XP_005173509.1 | 13 - 14 | 64 | |||
| XM_005173454.4 | XP_005173511.1 | 9 - 10 | 64 | |||
| GenBank mRNA | BC095683.1 | - | 13 - 14 | 1364 | 64 | |
More Information
Set Membership: |
|

cyb5r4
Genomic Map