Assay Details
| Gene Symbol: tlk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024951.2 | NP_001020122.2 | 4 - 5 | 596 | 121 | |
| XM_017356442.2 | XP_017211931.1 | 5 - 6 | 640 | 121 | ||
| GenBank mRNA | AF265345.1 | - | 4 - 5 | 561 | 121 | |
| BC071299.1 | - | 4 - 5 | 586 | 121 | ||
More Information
Set Membership: |
|

tlk1b
Genomic Map