Assay Details
| Gene Symbol: iah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025469.1 | NP_001020640.1 | 4 - 5 | 484 | 64 | |
| GenBank mRNA | BC095279.1 | - | 4 - 5 | 484 | 64 | |
| BC171513.1 | - | 4 - 5 | 473 | 64 | ||
| BC171519.1 | - | 4 - 5 | 473 | 64 | ||
More Information
Set Membership: |
|

iah1
Genomic Map