Assay Details
| Gene Symbol: emp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025508.1 | NP_001020679.1 | 2 - 3 | 302 | 60 | |
| GenBank mRNA | BC097185.1 | - | 2 - 3 | 302 | 60 | |
More Information
Set Membership: |
|

emp3b
Genomic Map