Assay Details
| Gene Symbol: szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030076.2 | NP_001025247.1 | 3 - 4 | 423 | 68 | |
| GenBank mRNA | BC076300.1 | - | 3 - 4 | 425 | 68 | |
| BC095055.1 | - | 3 - 4 | 424 | 68 | ||
More Information
Set Membership: |
|

szrd1
Genomic Map