Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 13 - 14 | 1606 | 111 | |
| XM_005170181.4 | XP_005170238.1 | 14 - 15 | 1707 | 111 | ||
| XM_021468448.1 | XP_021324123.1 | 15 - 16 | 1743 | 111 | ||
| XM_021468449.1 | XP_021324124.1 | 14 - 15 | 1662 | 111 | ||
| GenBank mRNA | BC096861.1 | - | 13 - 14 | 1550 | 111 | |
| BC115194.1 | - | 15 - 16 | 1772 | 111 | ||
| BC163586.1 | - | 13 - 14 | 1425 | 111 | ||
| BC163590.1 | - | 13 - 14 | 1425 | 111 | ||
| BC167965.1 | - | 13 - 14 | 1587 | 111 | ||
| BE017115.1 | - | 2 - 3 | 176 | 111 | ||
| CK677504.1 | - | 1 - 2 | 81 | 111 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map