Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 15 - 16 | 1765 | 101 | |
| XM_005170181.4 | XP_005170238.1 | 16 - 17 | 1866 | 101 | ||
| XM_021468448.1 | XP_021324123.1 | 17 - 18 | 1902 | 101 | ||
| XM_021468449.1 | XP_021324124.1 | 16 - 17 | 1821 | 101 | ||
| GenBank mRNA | BC096861.1 | - | 15 - 16 | 1709 | 101 | |
| BC115194.1 | - | 17 - 18 | 1931 | 101 | ||
| BC163586.1 | - | 15 - 16 | 1584 | 101 | ||
| BC163590.1 | - | 15 - 16 | 1584 | 101 | ||
| BC167965.1 | - | 15 - 16 | 1746 | 101 | ||
| BE017115.1 | - | 4 - 5 | 335 | 101 | ||
| CK677504.1 | - | 3 - 4 | 240 | 101 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map