Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 16 - 17 | 1886 | 66 | |
| XM_005170181.4 | XP_005170238.1 | 17 - 18 | 1987 | 66 | ||
| XM_021468448.1 | XP_021324123.1 | 18 - 19 | 2023 | 66 | ||
| XM_021468449.1 | XP_021324124.1 | 17 - 18 | 1942 | 66 | ||
| GenBank mRNA | BC096861.1 | - | 16 - 17 | 1830 | 66 | |
| BC115194.1 | - | 18 - 19 | 2052 | 66 | ||
| BC163586.1 | - | 16 - 17 | 1705 | 66 | ||
| BC163590.1 | - | 16 - 17 | 1705 | 66 | ||
| BC167965.1 | - | 16 - 17 | 1867 | 66 | ||
| BE017115.1 | - | 5 - 6 | 456 | 66 | ||
| CK677504.1 | - | 4 - 5 | 361 | 66 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map