Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 6 - 7 | 852 | 70 | |
| XM_005170181.4 | XP_005170238.1 | 7 - 8 | 953 | 70 | ||
| XM_021468448.1 | XP_021324123.1 | 8 - 9 | 989 | 70 | ||
| XM_021468449.1 | XP_021324124.1 | 7 - 8 | 908 | 70 | ||
| GenBank mRNA | BC096861.1 | - | 6 - 7 | 796 | 70 | |
| BC115194.1 | - | 7 - 8 | 936 | 70 | ||
| BC163586.1 | - | 6 - 7 | 671 | 70 | ||
| BC163590.1 | - | 6 - 7 | 671 | 70 | ||
| BC167965.1 | - | 6 - 7 | 833 | 70 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map