Assay Details
| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 8 - 9 | 1068 | 91 | |
| XM_005170181.4 | XP_005170238.1 | 9 - 10 | 1169 | 91 | ||
| XM_021468448.1 | XP_021324123.1 | 10 - 11 | 1205 | 91 | ||
| XM_021468449.1 | XP_021324124.1 | 9 - 10 | 1124 | 91 | ||
| GenBank mRNA | BC096861.1 | - | 8 - 9 | 1012 | 91 | |
| BC115194.1 | - | 10 - 11 | 1234 | 91 | ||
| BC163586.1 | - | 8 - 9 | 887 | 91 | ||
| BC163590.1 | - | 8 - 9 | 887 | 91 | ||
| BC167965.1 | - | 8 - 9 | 1049 | 91 | ||
More Information
Set Membership: |
|

snx9b
Genomic Map