Assay Details
| Gene Symbol: vps37bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030214.1 | NP_001025385.1 | 3 - 4 | 372 | 76 | |
| GenBank mRNA | BC097222.1 | - | 3 - 4 | 372 | 76 | |
More Information
Set Membership: |
|

vps37bb
Genomic Map