Assay Details
| Gene Symbol: blk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030220.1 | NP_001025391.1 | 13 - 14 | 1347 | 75 | |
| XM_017352511.2 | XP_017208000.1 | 12 - 13 | 1947 | 75 | ||
More Information
Set Membership: |
|

blk
Genomic Map