Assay Details
| Gene Symbol: atp2a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030277.1 | NP_001025448.1 | 12 - 13 | 1760 | 57 | |
| XM_005161196.4 | XP_005161253.1 | 13 - 14 | 2288 | 57 | ||
More Information
Set Membership: |
|

atp2a2b
Genomic Map