Assay Details
| Gene Symbol: pora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034982.1 | NP_001030154.1 | 10 - 11 | 1113 | 105 | |
| XM_005172678.4 | XP_005172735.1 | 9 - 10 | 1131 | 105 | ||
| GenBank mRNA | BC099998.1 | - | 10 - 11 | 1113 | 105 | |
More Information
Set Membership: |
|

pora
Genomic Map