Assay Details
| Gene Symbol: ca10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037121.1 | NP_001032198.1 | 4 - 5 | 592 | 94 | |
| GenBank mRNA | BC107833.1 | - | 4 - 5 | 592 | 94 | |
More Information
Set Membership: |
|

ca10a
Genomic Map