Assay Details
| Gene Symbol: ro60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037397.1 | NP_001032474.1 | 4 - 5 | 1285 | 78 | |
| GenBank mRNA | BC107964.1 | - | 4 - 5 | 1285 | 78 | |
| BC153401.1 | - | 3 - 4 | 485 | 78 | ||
More Information
Set Membership: |
|

ro60
Genomic Map