Assay Details
| Gene Symbol: bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039107.1 | NP_001034196.1 | 4 - 5 | 370 | 61 | |
| GenBank mRNA | AB212760.1 | - | 4 - 5 | 370 | 61 | |
| BC163162.1 | - | 4 - 5 | 370 | 61 | ||
| BC163191.1 | - | 4 - 5 | 370 | 61 | ||
| DQ779789.1 | - | 4 - 5 | 430 | 61 | ||
More Information
Set Membership: |
|

bhlhe41
Genomic Map