Assay Details
| Gene Symbol: tigara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039836.1 | NP_001034925.1 | 4 - 5 | 433 | 96 | |
| XM_021470524.1 | XP_021326199.1 | 3 - 4 | 156 | 96 | ||
| XM_021470525.1 | XP_021326200.1 | 2 - 3 | 191 | 96 | ||
| GenBank mRNA | BC114302.1 | - | 4 - 5 | 433 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021470525.1 transcripts.
Set Membership: |
|

tigara
Genomic Map