Assay Details
| Gene Symbol: olah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039932.1 | NP_001035021.1 | 2 - 3 | 436 | 77 | |
| XM_005159455.3 | XP_005159512.1 | 2 - 3 | 218 | 77 | ||
| XM_021468123.1 | XP_021323798.1 | 2 - 3 | 440 | 77 | ||
| GenBank mRNA | BC114310.1 | - | 2 - 3 | 436 | 77 | |
More Information
Set Membership: |
|

olah
Genomic Map