Assay Details
| Gene Symbol: cpox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040094.2 | NP_001035183.2 | 4 - 5 | 978 | 91 | |
| GenBank mRNA | AB180841.1 | - | 4 - 5 | 990 | 91 | |
| BC124459.1 | - | 4 - 5 | 978 | 91 | ||
More Information
Set Membership: |
|

cpox
Genomic Map