Assay Details
| Gene Symbol: gpat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040249.2 | NP_001035339.2 | 2 - 3 | 618 | 75 | |
| XM_021479195.1 | XP_021334870.1 | 2 - 3 | 560 | 75 | ||
| GenBank mRNA | BC115303.1 | - | 1 - 2 | 333 | 75 | |
| BC124075.1 | - | 2 - 3 | 618 | 75 | ||
| BC164676.1 | - | 1 - 2 | 203 | 75 | ||
More Information
Set Membership: |
|

gpat4
Genomic Map