Assay Details
| Gene Symbol: zpax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040389.1 | NP_001035479.1 | 10 - 11 | 1409 | 69 | |
| GenBank mRNA | BC115150.1 | - | 10 - 11 | 1409 | 69 | |
| BC124757.1 | - | 7 - 8 | 1068 | 69 | ||
| BC129309.1 | - | 7 - 8 | 1094 | 69 | ||
| BC146721.1 | - | 7 - 8 | 1047 | 69 | ||
More Information
Set Membership: |
|

zpax1
Genomic Map