Assay Details
| Gene Symbol: zpax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040389.1 | NP_001035479.1 | 3 - 4 | 295 | 85 | |
| GenBank mRNA | BC115150.1 | - | 3 - 4 | 295 | 85 | |
| BC124071.1 | - | 652 | 85 | |||
More Information
Set Membership: |
|

zpax1
Genomic Map