Assay Details
| Gene Symbol: limk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042691.2 | NP_001036156.1 | 14 - 15 | 1645 | 76 | |
| XM_005169645.4 | XP_005169702.1 | 12 - 13 | 1551 | 76 | ||
| XM_005169646.4 | XP_005169703.1 | 13 - 14 | 1590 | 76 | ||
| GenBank mRNA | BC163263.1 | - | 14 - 15 | 1645 | 76 | |
| DQ679229.1 | - | 14 - 15 | 1645 | 76 | ||
More Information
Set Membership: |
|

limk1a
Genomic Map