Assay Details
| Gene Symbol: nptxrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044314.1 | NP_001037779.1 | 2 - 3 | 1051 | 112 | |
| GenBank mRNA | BC067657.1 | - | 2 - 3 | 925 | 112 | |
| BC109464.1 | - | 3 - 4 | 963 | 112 | ||
| BC131861.1 | - | 2 - 3 | 1051 | 112 | ||
| BC171437.1 | - | 2 - 3 | 1040 | 112 | ||
| BC171439.1 | - | 2 - 3 | 1040 | 112 | ||
More Information
Set Membership: |
|

nptxrb
Genomic Map