Assay Details
| Gene Symbol: fam228a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045186.2 | NP_001038651.2 | 3 - 4 | 369 | 74 | |
| XM_017352515.2 | XP_017208004.1 | 3 - 4 | 427 | 74 | ||
| XM_021468502.1 | XP_021324177.1 | 2 - 3 | 1492 | 74 | ||
| GenBank mRNA | BC135057.1 | - | 3 - 4 | 369 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017352515.2 transcripts.
Set Membership: |
|

fam228a
Genomic Map